(4R,4'R)-4,4'-Diisopropyl-4,4',5,5'-tetrahydro-2,2'-bioxazole ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1219904-100MG
A1219904-1G
A1219904-250MG
A1219904-5G
76678-582EA
25
USD
76678-582
76678-584
76678-586
76678-588
(4R,4'R)-4,4'-Diisopropyl-4,4',5,5'-tetrahydro-2,2'-bioxazole ≥97%, ee 99%
(4R,4'R)-4,4'-Diisopropyl-4,4',5,5'-tetrahydro-2,2'-bioxazole
Precaución: for Research Use Only
Formula:
C₁₂H₂₀N₂O₂ MW: 224.2994 g/mol |
MDL Number:
MFCD32639896 CAS Number: 148925-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...