2,6-Bis((4R,5R)-4,5-dihydro-4,5-diphenyl-2-oxazolyl)pyridine ≥97%, ee 99%

Supplier: Ambeed
Warning

A1219903-1G A1219903-250MG A1219903-100MG
76678-578EA 549 USD
76678-578 76678-580 77866-750
2,6-Bis((4R,5R)-4,5-dihydro-4,5-diphenyl-2-oxazolyl)pyridine ≥97%, ee 99%
2,6-Bis((4R,5R)-4,5-dihydro-4,5-diphenyl-2-oxazolyl)pyridine

Precaución: for Research Use Only

Formula: C₃₅H₂₇N₃O₂
MW: 521.6078 g/mol
CAS Number: 372200-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR