Bis(2-(di-tert-butylphosphino)ethyl)amine hydrochloride 97%

Supplier: Ambeed
Warning

A1219877-1G A1219877-250MG A1219877-5G
76678-572EA 108 USD
76678-572 76678-574 76678-576
Bis(2-(di-tert-butylphosphino)ethyl)amine hydrochloride 97%
Bis(2-(di-tert-butylphosphino)ethyl)amine hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₄₆ClNP₂
MDL Number: MFCD31720637
CAS Number: 1326805-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR