3,4,5,6-Tetrakis(3,6-di-tert-butyl-9H-carbazol-9-yl)phthalonitrile 97%

Supplier: Ambeed
Warning

A1219876-100MG A1219876-50MG
76678-568EA 103.97 USD
76678-568 76678-570
3,4,5,6-Tetrakis(3,6-di-tert-butyl-9H-carbazol-9-yl)phthalonitrile 97%
3,4,5,6-Tetrakis(3,6-di-tert-butyl-9H-carbazol-9-yl)phthalonitrile

Precaución: for Research Use Only

Formula: C₈₈H₉₆N₆
MW: 1237.7441 g/mol
CAS Number: 1469705-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR