3,4,5,6-Tetrakis(3,6-diphenyl-9H-carbazol-9-yl)phthalonitrile 98%

Supplier: Ambeed
Warning

A1219875-1G A1219875-50MG A1219875-250MG
76678-566EA 623.81 USD
76678-566 77596-408 77621-020
3,4,5,6-Tetrakis(3,6-diphenyl-9H-carbazol-9-yl)phthalonitrile 98%
3,4,5,6-Tetrakis(3,6-diphenyl-9H-carbazol-9-yl)phthalonitrile

Precaución: for Research Use Only

Formula: C₁₀₄H₆₄N₆
MW: 1397.6612 g/mol
CAS Number: 1469707-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR