10-Ethyl-3,7,8-trimethylbenzo[g]pteridine-2,4(3H,10H)-dione 97%

Supplier: Ambeed
Warning

A1219873-100MG A1219873-1G A1219873-250MG A1219873-50MG
76678-558EA 284.33 USD
76678-558 76678-560 76678-562 76678-564
10-Ethyl-3,7,8-trimethylbenzo[g]pteridine-2,4(3H,10H)-dione 97%
10-Ethyl-3,7,8-trimethylbenzo[g]pteridine-2,4(3H,10H)-dione

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₄O₂
MW: 284.3131 g/mol
MDL Number: MFCD30474981
CAS Number: 67767-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR