5,10-Di(naphthalen-2-yl)-5,10-dihydrophenazine 97%

Supplier: Ambeed
Warning

A1219872-100MG A1219872-250MG
76678-556EA 70 USD
76678-556 77621-018
5,10-Di(naphthalen-2-yl)-5,10-dihydrophenazine 97%
5,10-Di(naphthalen-2-yl)-5,10-dihydrophenazine

Precaución: for Research Use Only

Formula: C₃₂H₂₂N₂
MDL Number: MFCD31619182
CAS Number: 1934269-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR