3,7-Di([1,1'-biphenyl]-4-yl)-10-(naphthalen-1-yl)-10H-phenoxazine 95%

Supplier: Ambeed
Warning

Synonyms: Phenox O-PC A0202

A1219871-1G A1219871-250MG A1219871-50MG A1219871-100MG
76678-552EA 729.9 USD
76678-552 76678-554 77571-320 77596-406
3,7-Di([1,1'-biphenyl]-4-yl)-10-(naphthalen-1-yl)-10H-phenoxazine 95%
3,7-Di([1,1'-biphenyl]-4-yl)-10-(naphthalen-1-yl)-10H-phenoxazine

Precaución: for Research Use Only

Formula: C₄₆H₃₁NO
MW: 613.74 g/mol
MDL Number: MFCD31619181
CAS Number: 1987900-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR