2,4,6-Tris(diphenylamino)-5-fluoroisophthalonitrile 97%

Supplier: Ambeed
Warning

A1219870-100MG A1219870-1G A1219870-250MG
76678-546EA 49.87 USD
76678-546 76678-548 76678-550
2,4,6-Tris(diphenylamino)-5-fluoroisophthalonitrile 97%
2,4,6-Tris(diphenylamino)-5-fluoroisophthalonitrile

Precaución: for Research Use Only

Formula: C₄₄H₃₀FN₅
MW: 647.7409 g/mol
MDL Number: MFCD32062492
CAS Number: 2260543-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR