2,4,6-Tri(9H-carbazol-9-yl)-5-chloroisophthalonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3CzCIPN

A1219869-100MG A1219869-250MG A1219869-5G
76678-542EA 107.16 USD
76678-542 76678-544 77621-016
2,4,6-Tri(9H-carbazol-9-yl)-5-chloroisophthalonitrile 97%
2,4,6-Tri(9H-carbazol-9-yl)-5-chloroisophthalonitrile

Precaución: for Research Use Only

Formula: C₄₄H₂₄ClN₅
MW: 658.15 g/mol
MDL Number: MFCD32062494
CAS Number: 1469704-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR