2,4,6-Tris(diphenylamino)-3,5-difluorobenzonitrile 97%

Supplier: Ambeed
Warning

A1219868-100MG A1219868-1G A1219868-250MG A1219868-5G
76678-536EA 15 USD
76678-536 76678-538 76678-540 77621-014
2,4,6-Tris(diphenylamino)-3,5-difluorobenzonitrile 97%
2,4,6-Tris(diphenylamino)-3,5-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₄₃H₃₀F₂N₄
MDL Number: MFCD32062493
CAS Number: 1403850-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR