9-Mesityl-10-methylacridin-10-ium iodide 97%

Supplier: Ambeed
Warning

A1219867-100MG A1219867-1G A1219867-250MG
76678-530EA 24 USD
76678-530 76678-532 76678-534
9-Mesityl-10-methylacridin-10-ium iodide 97%
9-Mesityl-10-methylacridin-10-ium iodide

Precaución: for Research Use Only

Formula: C₂₃H₂₂IN
CAS Number: 930097-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR