10-(3,5-Dimethoxyphenyl)-9-mesityl-1,3,6,8-tetramethoxyacridin-10-ium tetrafluoroborate 97%

Supplier: Ambeed
Warning

A1219865-250MG A1219865-100MG A1219865-1G
76678-528EA 220.67 USD
76678-528 77571-316 77571-318
10-(3,5-Dimethoxyphenyl)-9-mesityl-1,3,6,8-tetramethoxyacridin-10-ium tetrafluoroborate 97%
10-(3,5-Dimethoxyphenyl)-9-mesityl-1,3,6,8-tetramethoxyacridin-10-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₃₄H₃₆BF₄NO₆
MW: 641.4574 g/mol
MDL Number: MFCD31381913
CAS Number: 1965330-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR