6,6'-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine 97%

Supplier: Ambeed
Warning

A1219863-100MG A1219863-1G A1219863-250MG
76678-522EA 60 USD
76678-522 76678-524 76678-526
6,6'-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine 97%
6,6'-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₂₄H₃₀N₄O₂
MW: 406.5206 g/mol
CAS Number: 2093382-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR