6,6'-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine ≥95%, ee 99%

Supplier: Ambeed
Warning

A1219862-100MG A1219862-1G A1219862-250MG
76678-516EA 96.55 USD
76678-516 76678-518 76678-520
6,6'-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine ≥95%, ee 99%
6,6'-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₃₀H₂₆N₄O₂
MW: 474.55 g/mol
MDL Number: MFCD00009593
CAS Number: 2088982-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR