6,6'-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine ≥97%, ee 99%

Supplier: Ambeed
Warning

A1219861-100MG A1219861-1G A1219861-250MG
76678-510EA 87 USD
76678-510 76678-512 76678-514
6,6'-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine ≥97%, ee 99%
6,6'-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₂₈H₂₂N₄O₂
MW: 446.4999 g/mol
CAS Number: 273216-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR