(E)-(2-(3-Methoxy-3-oxoprop-1-en-1-yl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A1219784-100MG A1219784-1G A1219784-250MG
76678-504EA 15 USD
76678-504 76678-506 76678-508
(E)-(2-(3-Methoxy-3-oxoprop-1-en-1-yl)phenyl)boronic acid 97%
(E)-(2-(3-Methoxy-3-oxoprop-1-en-1-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₄
MW: 206.0029 g/mol
MDL Number: MFCD02179475
CAS Number: 372193-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR