4-(trans-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (E)-(4-(3-ethoxy-3-oxoprop-1-en-1-yl)phenyl)boronic acid

A1219782-1G A1219782-5G
76678-500EA 56 USD
76678-500 76678-502
4-(trans-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid 98%
4-(trans-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BO₄
MDL Number: MFCD04115651
CAS Number: 850568-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR