4-Fluoro-3-(((5-(1-methyl-1H-pyrazol-3-yl)pyridin-3-yl)methyl)amino)-N-(3-(trifluoromethoxy)phenyl)benzamide 97%

Supplier: Ambeed
Warning

A1219698-1MG A1219698-25MG A1219698-50MG A1219698-5MG
76678-492EA 16 USD
76678-492 76678-494 76678-496 76678-498
4-Fluoro-3-(((5-(1-methyl-1H-pyrazol-3-yl)pyridin-3-yl)methyl)amino)-N-(3-(trifluoromethoxy)phenyl)benzamide 97%
4-Fluoro-3-(((5-(1-methyl-1H-pyrazol-3-yl)pyridin-3-yl)methyl)amino)-N-(3-(trifluoromethoxy)phenyl)benzamide

Precaución: for Research Use Only

Formula: C₂₄H₁₉F₄N₅O₂
CAS Number: 2442597-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR