2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)acetic acid 98%

Supplier: Ambeed
Danger

Synonyms: Thalidomide-NH-CH2-COOH

A1219680-100MG A1219680-1G A1219680-250MG A1219680-5G
76678-484EA 55.17 USD
76678-484 76678-486 76678-488 77621-012
2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)acetic acid 98%
2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)acetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃N₃O₆
MW: 331.28 g/mol
MDL Number: MFCD31667076
CAS Number: 927670-97-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR