Pyrylium tetrafluoroborate 97%

Supplier: Ambeed
Warning

A1219676-250MG A1219676-100MG
76678-482EA 415.88 USD
76678-482 77566-280
Pyrylium tetrafluoroborate 97%
Pyrylium tetrafluoroborate

Precaución: For research use only.

Formula: C₅H₅BF₄O
MW: 167.897 g/mol
MDL Number: MFCD32857267
CAS Number: 80279-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR