6-Bromo-4-(6-fluoropyridin-3-yl)pyrazolo[1,5-a]pyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A1219492-100MG A1219492-1G A1219492-250MG A1219492-5G
76678-468EA 72.15 USD
76678-468 76678-470 76678-472 77566-278
6-Bromo-4-(6-fluoropyridin-3-yl)pyrazolo[1,5-a]pyridine-3-carbonitrile 97%
6-Bromo-4-(6-fluoropyridin-3-yl)pyrazolo[1,5-a]pyridine-3-carbonitrile

Precaución: For research use only.

Formula: C₁₃H₆BrFN₄
MW: 317.1159 g/mol
MDL Number: MFCD31697791
CAS Number: 2222756-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR