2-Ethylbutyl ((S)-(perfluorophenoxy)(phenoxy)phosphoryl)-L-alaninate 98%

Supplier: Ambeed
Warning

A1219324-1G A1219324-25G A1219324-5G
76678-454EA 7 USD
76678-454 76678-456 76678-458
2-Ethylbutyl ((S)-(perfluorophenoxy)(phenoxy)phosphoryl)-L-alaninate 98%
2-Ethylbutyl ((S)-(perfluorophenoxy)(phenoxy)phosphoryl)-L-alaninate

Precaución: for Research Use Only

Formula: C₂₁H₂₃F₅NO₅P
MDL Number: MFCD30803468
CAS Number: 1911578-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR