(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(pent-4-ynamido)hexanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Lys(4-Pentynoyl)-OH

A1219288-100MG A1219288-1G A1219288-250MG
76678-448EA 40.32 USD
76678-448 76678-450 76678-452
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(pent-4-ynamido)hexanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(pent-4-ynamido)hexanoic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₈N₂O₅
MW: 448.5109 g/mol
MDL Number: MFCD31380695
CAS Number: 1159531-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR