(4-Benzhydrylpiperazin-1-yl)(1,5-dimethyl-1H-pyrazol-3-yl)methanone 98%

Supplier: Ambeed
Warning

A1219206-10MG A1219206-1MG A1219206-25MG A1219206-50MG A1219206-5MG
76678-416EA 46 USD
76678-416 76678-418 76678-420 76678-422 76678-424
(4-Benzhydrylpiperazin-1-yl)(1,5-dimethyl-1H-pyrazol-3-yl)methanone 98%
(4-Benzhydrylpiperazin-1-yl)(1,5-dimethyl-1H-pyrazol-3-yl)methanone

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₄O
MDL Number: MFCD04123707
CAS Number: 958323-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR