2-(3-Chloro-4-fluorophenyl)-N-(1-cyclohexyl-3-methyl-1H-pyrazol-5-yl)acetamide 98%

Supplier: Ambeed
Warning

Synonyms: VU0810464

A1219204-10MG A1219204-1MG A1219204-25MG A1219204-50MG A1219204-5MG
76678-406EA 30.77 USD
76678-406 76678-408 76678-410 76678-412 76678-414
2-(3-Chloro-4-fluorophenyl)-N-(1-cyclohexyl-3-methyl-1H-pyrazol-5-yl)acetamide 98%
2-(3-Chloro-4-fluorophenyl)-N-(1-cyclohexyl-3-methyl-1H-pyrazol-5-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₈H₂₁ClFN₃O
MW: 349.83 g/mol
CAS Number: 2126040-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR