2-((5-Chloroquinolin-8-yl)oxy)-N,N-diethylethanamine dihydrochloride 98%

Supplier: Ambeed
Warning

A1219196-10MG A1219196-1MG A1219196-25MG A1219196-50MG A1219196-5MG
76678-396EA 33 USD
76678-396 76678-398 76678-400 76678-402 76678-404
2-((5-Chloroquinolin-8-yl)oxy)-N,N-diethylethanamine dihydrochloride 98%
2-((5-Chloroquinolin-8-yl)oxy)-N,N-diethylethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₁Cl₃N₂O
MDL Number: MFCD32667019
CAS Number: 861038-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR