2-Allyl-6-(((1-propyl-1H-benzo[d]imidazol-2-yl)amino)methyl)phenol 98%

Supplier: Ambeed
Warning

Synonyms: CM10

A1219192-100MG A1219192-10MG A1219192-1MG A1219192-25MG A1219192-50MG A1219192-5MG
76678-374EA 250.38 USD
76678-374 76678-376 76678-378 76678-380 76678-382 76678-384
2-Allyl-6-(((1-propyl-1H-benzo[d]imidazol-2-yl)amino)methyl)phenol 98%
2-Allyl-6-(((1-propyl-1H-benzo[d]imidazol-2-yl)amino)methyl)phenol

Precaución: for Research Use Only

Formula: C₂₀H₂₃N₃O
MW: 321.42 g/mol
MDL Number: MFCD03683082
CAS Number: 692269-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR