(E)-1,2-Bis(4-(1H-imidazol-1-yl)phenyl)-1,2-diphenylethene 97%

Supplier: Ambeed
Warning

A1219152-1G A1219152-250MG
76678-348EA 63.66 USD
76678-348 76678-350
(E)-1,2-Bis(4-(1H-imidazol-1-yl)phenyl)-1,2-diphenylethene 97%
(E)-1,2-Bis(4-(1H-imidazol-1-yl)phenyl)-1,2-diphenylethene

Precaución: for Research Use Only

Formula: C₃₂H₂₄N₄
MW: 464.5598 g/mol
CAS Number: 2828431-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR