2-Methoxyethyl 2-cyanoacrylate 98%

Supplier: Ambeed
Warning

A1219013-5G A1219013-100G
76678-344EA 9 USD
76678-344 77571-304
2-Methoxyethyl 2-cyanoacrylate 98%
2-Methoxyethyl 2-cyanoacrylate

Precaución: for Research Use Only

Formula: C₇H₉NO₃
MDL Number: MFCD28361936
CAS Number: 27816-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR