N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-(piperazin-1-yl)propoxy)quinazolin-4-amine 97%

Supplier: Ambeed
Warning

A1218794-250MG A1218794-1G A1218794-100MG
76678-342EA 374 USD
76678-342 77571-298 77866-728
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-(piperazin-1-yl)propoxy)quinazolin-4-amine 97%
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-(piperazin-1-yl)propoxy)quinazolin-4-amine

Precaución: for Research Use Only

Formula: C₂₂H₂₅ClFN₅O₂
MW: 445.9176 g/mol
CAS Number: 295330-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR