4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)bicyclo[1.1.1]pentan-1-yl)-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1218000-25MG A1218000-50MG A1218000-100MG
76678-324EA 145.35 USD
76678-324 76678-326 77620-974
4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)bicyclo[1.1.1]pentan-1-yl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)bicyclo[1.1.1]pentan-1-yl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₈BF₃O₂
MW: 262.0763 g/mol
MDL Number: MFCD30181078
CAS Number: 2410440-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR