2-(Pyrrolidin-3-yl)pyridine dihydrochloride 95%

Supplier: Ambeed
Warning

A1217988-100MG A1217988-250MG
76678-320EA 61 USD
76678-320 76678-322
2-(Pyrrolidin-3-yl)pyridine dihydrochloride 95%
2-(Pyrrolidin-3-yl)pyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄Cl₂N₂
MDL Number: MFCD31446554
CAS Number: 2138411-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR