2-[3'-Bromo[1,1'-biphenyl]-3-yl]-4,6-diphenyl-1,3,5-triazine ≥98%
Supplier: Ambeed
|
Warning
|
A1217319-1G
A1217319-5G
76678-310EA
7
USD
76678-310
76678-312
2-[3'-Bromo[1,1'-biphenyl]-3-yl]-4,6-diphenyl-1,3,5-triazine ≥98%
2-[3'-Bromo[1,1'-biphenyl]-3-yl]-4,6-diphenyl-1,3,5-triazine
Precaución: for Research Use Only
Formula:
C₂₇H₁₈BrN₃ |
MDL Number:
MFCD30730127 CAS Number: 1606981-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...