(S)-7-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride 97%

Supplier: Ambeed
Warning

A1217089-100MG A1217089-250MG
76678-304EA 95 USD
76678-304 76678-306
(S)-7-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride 97%
(S)-7-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉ClFNO
MDL Number: MFCD24435422
CAS Number: 2453296-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR