(E)-2-(2,3-Dichlorobenzylidene)hydrazine-1-carboximidamide 98%

Supplier: Ambeed
Warning

A1216819-10MG A1216819-1MG A1216819-25MG A1216819-5MG
76678-296EA 44 USD
76678-296 76678-298 76678-300 76678-302
(E)-2-(2,3-Dichlorobenzylidene)hydrazine-1-carboximidamide 98%
(E)-2-(2,3-Dichlorobenzylidene)hydrazine-1-carboximidamide

Precaución: for Research Use Only

Formula: C₈H₈Cl₂N₄
MDL Number: MFCD03653280
CAS Number: 2022961-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR