N-(3-Methoxybenzyl)-5-(4-methoxyphenyl)-1-methyl-1H-imidazol-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: MALAT1-IN-1

A1216706-10MG A1216706-1MG A1216706-25MG A1216706-50MG A1216706-5MG
76678-286EA 43.5 USD
76678-286 76678-288 76678-290 76678-292 76678-294
N-(3-Methoxybenzyl)-5-(4-methoxyphenyl)-1-methyl-1H-imidazol-2-amine 98%
N-(3-Methoxybenzyl)-5-(4-methoxyphenyl)-1-methyl-1H-imidazol-2-amine

Precaución: for Research Use Only

Formula: C₁₉H₂₁N₃O₂
MW: 323.39 g/mol
MDL Number: MFCD06011226
CAS Number: 827327-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR