1-(7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl)-2-phenylethane-1,2-dione 98%

Supplier: Ambeed
Warning

Synonyms: IRAK4-IN-4

A1216693-10MG A1216693-1MG A1216693-25MG A1216693-50MG A1216693-5MG
76678-276EA 117.76 USD
76678-276 76678-278 76678-280 76678-282 76678-284
1-(7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl)-2-phenylethane-1,2-dione 98%
1-(7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl)-2-phenylethane-1,2-dione

Precaución: for Research Use Only

Formula: C₂₂H₁₆N₂O₂
MW: 340.3747 g/mol
CAS Number: 1850276-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR