(S)-2,2'-diiodo-1,1'-binaphthalene 95+%

Supplier: Ambeed
Danger

A1216580-250MG A1216580-100MG
76678-274EA 58 USD
76678-274 77866-708
(S)-2,2'-diiodo-1,1'-binaphthalene 95+%
(S)-2,2'-diiodo-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₂₀H₁₂I₂
MW: 506.1200 g/mol
CAS Number: 86688-07-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR