(S)-4-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride 98%

Supplier: Ambeed
Warning

A1216492-100MG A1216492-250MG
76678-270EA 81 USD
76678-270 76678-272
(S)-4-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride 98%
(S)-4-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉ClFNO
CAS Number: 2177263-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR