Methyl 2-bromo-4-chloro-6-fluorobenzoate 98%

Supplier: Ambeed
Warning

A1215516-250MG A1215516-5G A1215516-1G
76678-268EA 11 USD
76678-268 77571-284 77866-706
Methyl 2-bromo-4-chloro-6-fluorobenzoate 98%
Methyl 2-bromo-4-chloro-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
MDL Number: MFCD28733924
CAS Number: 943975-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR