2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindoline-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A1215373-100MG A1215373-1G A1215373-250MG A1215373-5G A1215373-25G
76678-260EA 23 USD
76678-260 76678-262 76678-264 77592-368 77620-940
2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindoline-5-carboxylic acid 98%
2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindoline-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O₆
MDL Number: MFCD32063447
CAS Number: 1216805-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR