5-(Bromomethyl)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%

Supplier: Ambeed
Danger

A1215370-1G A1215370-250MG A1215370-5G
76678-258EA 320.4 USD
76678-258 77596-354 77620-938
5-(Bromomethyl)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%
5-(Bromomethyl)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂O₄
MW: 351.1521 g/mol
MDL Number: MFCD34601661
CAS Number: 1312023-72-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR