4-Chloro-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%

Supplier: Ambeed
Warning

A1215364-100MG A1215364-5G
76678-252EA 18 USD
76678-252 76678-254
4-Chloro-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%
4-Chloro-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₃H₉ClN₂O₄
MDL Number: MFCD30188071
CAS Number: 244057-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR