4-Bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%

Supplier: Ambeed
Warning

A1215363-1G A1215363-250MG A1215363-25G A1215363-5G
76678-244EA 22 USD
76678-244 76678-246 76678-248 76678-250
4-Bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%
4-Bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂O₄
MDL Number: MFCD31730866
CAS Number: 2093536-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR