Bis(2-ethylhexyl)terephthalate 98%

Supplier: Ambeed
Warning

A1215261-1000G A1215261-100G A1215261-25G A1215261-500G
76678-226EA 33 USD
76678-226 76678-228 76678-230 76678-232
Bis(2-ethylhexyl)terephthalate 98%
Bis(2-ethylhexyl)terephthalate

Precaución: for Research Use Only

Formula: C₂₄H₃₈O₄
MDL Number: MFCD00072256
CAS Number: 6422-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR