1-(4-Chlorophenyl)-3-(1-methyl-1H-pyrazol-4-yl)propane-1,3-dione 97%

Supplier: Ambeed
Warning

A1215131-100MG A1215131-250MG
76678-222EA 66.84 USD
76678-222 76678-224
1-(4-Chlorophenyl)-3-(1-methyl-1H-pyrazol-4-yl)propane-1,3-dione 97%
1-(4-Chlorophenyl)-3-(1-methyl-1H-pyrazol-4-yl)propane-1,3-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClN₂O₂
MW: 262.6916 g/mol
MDL Number: MFCD25155745
CAS Number: 1503529-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR