6-Bromo-7-fluoro-3,4-dihydronaphthalen-1(2H)-one 97%

Supplier: Ambeed
Warning

A1215123-100MG A1215123-1G A1215123-250MG
76678-214EA 99.73 USD
76678-214 76678-216 76678-218
6-Bromo-7-fluoro-3,4-dihydronaphthalen-1(2H)-one 97%
6-Bromo-7-fluoro-3,4-dihydronaphthalen-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₈BrFO
MW: 243.0723 g/mol
MDL Number: MFCD18208386
CAS Number: 1260014-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR