9'-Phenyl-9H,9'H-2,3'-bicarbazole 98%

Supplier: Ambeed
Warning

A1214199-1G A1214199-250MG
76678-200EA 50.93 USD
76678-200 76678-202
9'-Phenyl-9H,9'H-2,3'-bicarbazole 98%
9'-Phenyl-9H,9'H-2,3'-bicarbazole

Precaución: for Research Use Only

Formula: C₃₀H₂₀N₂
MW: 408.4932 g/mol
MDL Number: MFCD32012082
CAS Number: 1345202-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR