2,3-Dibromo-9H-carbazole 98%

Supplier: Ambeed
Warning

A1214194-100MG A1214194-250MG A1214194-1G
76678-196EA 44 USD
76678-196 76678-198 77620-930
2,3-Dibromo-9H-carbazole 98%
2,3-Dibromo-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₂H₇Br₂N
MW: 324.9987 g/mol
CAS Number: 1356059-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR